tert-Butyl 3,4-dihydro-1H-pyrido[3,4-b]indole-2(9H)-carboxylate 95%

Supplier: Ambeed
Warning

A664204-1G A664204-250MG A664204-5G
77104-594EA 55.17 USD
77104-594 77104-596 77104-598
tert-Butyl 3,4-dihydro-1H-pyrido[3,4-b]indole-2(9H)-carboxylate 95%
tert-Butyl 3,4-dihydro-1H-pyrido[3,4-b]indole-2(9H)-carboxylate

Precaución: for Research Use Only

Formula: C₁₆H₂₀N₂O₂
MW: 272.34 g/mol
MDL Number: MFCD09029520
CAS Number: 168824-94-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR