Cyclopropyl(4-hydroxyphenyl)methanone 98%

Supplier: Ambeed
Warning

A662230-100MG A662230-250MG A662230-5G
77104-586EA 7 USD
77104-586 77104-588 77104-590
Cyclopropyl(4-hydroxyphenyl)methanone 98%
Cyclopropyl(4-hydroxyphenyl)methanone

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₂
MDL Number: MFCD06200217
CAS Number: 36116-18-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR