Potassium-2-fluoro-4-(trifluoromethyl)phenyltrifluoroborate 96%
Supplier: Ambeed
|
Warning
|
Synonyms:
Potassium 2-fluoro-4-(trifluoromethyl)phenyltrifluoroborate
A661730-250MG
A661730-5G
77104-582EA
18.04
USD
77104-582
77104-584
Potassium-2-fluoro-4-(trifluoromethyl)phenyltrifluoroborate 96%
Potassium 2-fluoro-4-(trifluoromethyl)phenyltrifluoroborate
Precaución: for Research Use Only
Formula:
C₇H₃BF₇K MW: 269.9969 g/mol |
MDL Number:
MFCD07781236 CAS Number: 1150655-12-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...