4-Amino-1-methylpyrazole-3-carboxylic acid 95%

Supplier: Ambeed
Warning

A659536-100MG A659536-1G A659536-250MG
77104-568EA 51.99 USD
77104-568 77104-570 77104-572
4-Amino-1-methylpyrazole-3-carboxylic acid 95%
4-Amino-1-methylpyrazole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₇N₃O₂
MW: 141.128 g/mol
MDL Number: MFCD08558492
CAS Number: 1006334-34-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR