3,4-Dichloro-5-fluorobenzoic acid 95+%

Supplier: Ambeed
Warning

A659511-100MG A659511-250MG
77104-564EA 50.93 USD
77104-564 77104-566
3,4-Dichloro-5-fluorobenzoic acid 95+%
3,4-Dichloro-5-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃Cl₂FO₂
MW: 209.0019 g/mol
MDL Number: MFCD11845916
CAS Number: 1160574-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR