(3-Chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanol 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Chloro-3-(hydroxymethyl)phenylboronic acid pinacol ester
A657949-1G
A657949-250MG
A657949-5G
77104-550EA
209
USD
77104-550
77104-552
77104-554
(3-Chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanol 98%
(3-Chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanol
Precaución: for Research Use Only
Formula:
C₁₃H₁₈BClO₃ MW: 268.54 g/mol |
MDL Number:
MFCD18374004 CAS Number: 1112210-59-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...