2,6-Dichlorobenzyl alcohol 95%

Supplier: Ambeed
Warning

Synonyms: (2,6-Dichlorophenyl)methanol

A657108-100G A657108-500G A657108-5G A657108-25G
77104-546EA 83.82 USD
77104-546 77104-548 77614-028 77686-626
2,6-Dichlorobenzyl alcohol 95%
2,6-Dichlorobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₆Cl₂O
MW: 177.03 g/mol
MDL Number: MFCD00004609
CAS Number: 15258-73-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR