2-(2-Chloro-4-nitrophenyl)acetic acid 95%

Supplier: Ambeed
Warning

A655869-1G A655869-250MG
77104-534EA 90 USD
77104-534 77104-536
2-(2-Chloro-4-nitrophenyl)acetic acid 95%
2-(2-Chloro-4-nitrophenyl)acetic acid

Precaución: for Research Use Only

Formula: C₈H₆ClNO₄
MDL Number: MFCD11036396
CAS Number: 73088-11-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR