2-(4-Methylphenoxy)benzylamine hydrochloride 95%

Supplier: Ambeed
Danger

Synonyms: (2-(p-Tolyloxy)phenyl)methanamine hydrochloride

A655146-100MG A655146-1G A655146-250MG A655146-5G
77104-526EA 23.34 USD
77104-526 77104-528 77104-530 77104-532
2-(4-Methylphenoxy)benzylamine hydrochloride 95%
2-(4-Methylphenoxy)benzylamine hydrochloride

Precaución: for Research Use Only

Formula: C₁₄H₁₅NO·ClH
MW: 249.736 g/mol
MDL Number: MFCD03840021
CAS Number: 1171318-10-7
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR