Potassium (1-(tert-butoxycarbonyl)-1H-indol-3-yl)trifluoroborate 96%

Supplier: Ambeed
Warning

A654662-1G A654662-250MG A654662-5G
77104-520EA 207 USD
77104-520 77104-522 77104-524
Potassium (1-(tert-butoxycarbonyl)-1H-indol-3-yl)trifluoroborate 96%
Potassium (1-(tert-butoxycarbonyl)-1H-indol-3-yl)trifluoroborate

Precaución: for Research Use Only

Formula: C₁₃H₁₄BF₃KNO₂
MDL Number: MFCD28014052
CAS Number: 1428884-69-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR