1-(Pyrimidin-2-yl)piperidine-4-carboxylic acid 98%

Supplier: Ambeed
Warning

A653777-1G A653777-250MG A653777-5G
77104-514EA 44.56 USD
77104-514 77104-516 77104-518
1-(Pyrimidin-2-yl)piperidine-4-carboxylic acid 98%
1-(Pyrimidin-2-yl)piperidine-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₃N₃O₂
MW: 207.23 g/mol
MDL Number: MFCD00664474
CAS Number: 303144-44-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR