3-(Methylsulfonyl)benzaldehyde 95%

Supplier: Ambeed
Warning

Synonyms: 3-Formylphenyl methyl sulphone

A652509-100MG A652509-1G
77104-510EA 9 USD
77104-510 77104-512
3-(Methylsulfonyl)benzaldehyde 95%
3-(Methylsulfonyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₈O₃S
MDL Number: MFCD11052329
CAS Number: 43114-43-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR