Bromocyclopropane 98%

Supplier: Ambeed
Danger

A652290-5G A652290-10G A652290-25G
77104-508EA 9 USD
77104-508 77551-976 77551-978
Bromocyclopropane 98%
Bromocyclopropane

Precaución: for Research Use Only

Formula: C₃H₅Br
MDL Number: MFCD00001271
CAS Number: 4333-56-6
UN: 1993
ADR: 3,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR