Ethyl 5-amino-1-methyl-1H-imidazole-4-carboxylate 95%

Supplier: Ambeed
Warning

A650747-100MG A650747-1G A650747-250MG A650747-5G
77104-498EA 54.11 USD
77104-498 77104-500 77104-502 77104-504
Ethyl 5-amino-1-methyl-1H-imidazole-4-carboxylate 95%
Ethyl 5-amino-1-methyl-1H-imidazole-4-carboxylate

Precaución: for Research Use Only

Formula: C₇H₁₁N₃O₂
MW: 169.18 g/mol
MDL Number: MFCD02239042
CAS Number: 54147-04-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR