Benzyl 6-bromospiro[indoline-3,4'-piperidine]-1'-carboxylate 95%

Supplier: Ambeed
Warning

A649372-100MG A649372-250MG
77104-494EA 418 USD
77104-494 77104-496
Benzyl 6-bromospiro[indoline-3,4'-piperidine]-1'-carboxylate 95%
Benzyl 6-bromospiro[indoline-3,4'-piperidine]-1'-carboxylate

Precaución: for Research Use Only

Formula: C₂₀H₂₁BrN₂O₂
MW: 401.30 g/mol
MDL Number: MFCD11975881
CAS Number: 473737-32-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR