Sodium 4-bromobenzenesulfinate dihydrate 98%
Supplier: Ambeed
|
Warning
|
A649295-1G
A649295-250MG
A649295-25G
A649295-5G
A649295-100G
77104-486EA
6
USD
77104-486
77104-488
77104-490
77104-492
77900-961
Sodium 4-bromobenzenesulfinate dihydrate 98%
Sodium 4-bromobenzenesulphinate dihydrate
Precaución: for Research Use Only
Formula:
C₆H₄BrNaO₂S·2H₂O |
MDL Number:
MFCD00210139 CAS Number: 175278-64-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...