Bis(2,5-dioxopyrrolidin-1-yl) 4,7,10,13-tetraoxahexadecane-1,16-dioate 98%

Supplier: Ambeed
Warning

A649050-1G A649050-100MG A649050-250MG A649050-5G
77104-484EA 109 USD
77104-484 77686-602 77686-603 77686-604
Bis(2,5-dioxopyrrolidin-1-yl) 4,7,10,13-tetraoxahexadecane-1,16-dioate 98%
Bis(2,5-dioxopyrrolidin-1-yl) 4,7,10,13-tetraoxahexadecane-1,16-dioate

Precaución: for Research Use Only

Formula: C₂₀H₂₈N₂O₁₂
MDL Number: MFCD20228963
CAS Number: 1314378-11-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR