(S)-3-Amino-1-benzylpiperidine 98%

Supplier: Ambeed
Warning

Synonyms: (S)-1-Benzylpiperidin-3-amine

A648613-1G A648613-5G
77104-480EA 41.38 USD
77104-480 77104-482
(S)-3-Amino-1-benzylpiperidine 98%
(S)-3-Amino-1-benzylpiperidine

Precaución: for Research Use Only

Formula: C₁₂H₁₈N₂
MW: 190.29 g/mol
MDL Number: MFCD03093375
CAS Number: 168466-85-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR