(4-Methoxyphenyl)acetyl chloride 98%

Supplier: Ambeed
Danger

A646126-1G A646126-25G A646126-5G
77104-460EA 15 USD
77104-460 77104-462 77104-464
(4-Methoxyphenyl)acetyl chloride 98%
(4-Methoxyphenyl)acetyl chloride

Precaución: for Research Use Only

Formula: C₉H₉ClO₂
MDL Number: MFCD00075474
CAS Number: 4693-91-8
UN: 3265
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR