7-Bromo-4-fluoro-1H-indazole 97%

Supplier: Ambeed
Warning

A645854-100MG A645854-1G A645854-250MG A645854-5G
77104-454EA 15 USD
77104-454 77104-456 77104-458 77686-599
7-Bromo-4-fluoro-1H-indazole 97%
7-Bromo-4-fluoro-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₄BrFN₂
MDL Number: MFCD09749914
CAS Number: 1000341-72-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR