4-(Piperidin-4-yl)benzonitrile hydrochloride 95%

Supplier: Ambeed
Warning

A645848-100MG A645848-1G A645848-250MG
77104-448EA 163.38 USD
77104-448 77104-450 77104-452
4-(Piperidin-4-yl)benzonitrile hydrochloride 95%
4-(Piperidin-4-yl)benzonitrile hydrochloride

Precaución: for Research Use Only

Formula: C₁₂H₁₄N₂·ClH
MW: 222.714 g/mol
MDL Number: MFCD08544124
CAS Number: 162997-34-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR