(1-(tert-Butoxycarbonyl)-1H-indazol-5-yl)boronic acid 95%

Supplier: Ambeed
Warning

A643828-100MG A643828-1G A643828-250MG A643828-5G
77104-438EA 78.51 USD
77104-438 77104-440 77104-442 77104-444
(1-(tert-Butoxycarbonyl)-1H-indazol-5-yl)boronic acid 95%
(1-(tert-Butoxycarbonyl)-1H-indazol-5-yl)boronic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₅BN₂O₄
MW: 262.06 g/mol
MDL Number: MFCD18837754
CAS Number: 1167418-12-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR