3-(3,5-Dimethyl-1H-pyrazol-1-yl)phenylboronic acid 98%

Supplier: Ambeed
Warning

A643442-100MG A643442-1G A643442-250MG A643442-5G
77104-430EA 37.14 USD
77104-430 77104-432 77104-434 77104-436
3-(3,5-Dimethyl-1H-pyrazol-1-yl)phenylboronic acid 98%
3-(3,5-Dimethyl-1H-pyrazol-1-yl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₃BN₂O₂
MW: 216.04 g/mol
MDL Number: MFCD08572151
CAS Number: 1025735-46-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR