(R)-3-Amino-1-benzylpiperidine 98%

Supplier: Ambeed
Danger

A642477-1G A642477-5G A642477-25G A642477-250MG
77104-426EA 22 USD
77104-426 77104-428 77551-948 77901-973
(R)-3-Amino-1-benzylpiperidine 98%
(R)-3-Amino-1-benzylpiperidine

Precaución: for Research Use Only

Formula: C₁₂H₁₈N₂
MDL Number: MFCD03093374
CAS Number: 168466-84-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR