9-(4-Nitrophenyl)carbazole 98%

Supplier: Ambeed
Warning

A642353-1G A642353-250MG A642353-5G
77104-420EA 35.01 USD
77104-420 77104-422 77104-424
9-(4-Nitrophenyl)carbazole 98%
9-(4-Nitrophenyl)carbazole

Precaución: for Research Use Only

Formula: C₁₈H₁₂N₂O₂
MW: 288.30 g/mol
MDL Number: MFCD00159886
CAS Number: 16982-76-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR