2'-Bromo-2,7-di-tert-butyl-9,9'-spirobi[fluorene] 98%

Supplier: Ambeed
Warning

A641064-1G A641064-250MG A641064-25G A641064-5G
77104-412EA 8.49 USD
77104-412 77104-414 77104-416 77104-418
2'-Bromo-2,7-di-tert-butyl-9,9'-spirobi[fluorene] 98%
2'-Bromo-2,7-di-tert-butyl-9,9'-spirobi[fluorene]

Precaución: for Research Use Only

Formula: C₃₃H₃₁Br
MW: 507.50 g/mol
MDL Number: MFCD20921691
CAS Number: 393841-81-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR