4-(4-Fluorophenoxy)piperidine 95%

Supplier: Ambeed
Warning

A640235-100MG A640235-1G A640235-250MG
77104-406EA 15 USD
77104-406 77104-408 77104-410
4-(4-Fluorophenoxy)piperidine 95%
4-(4-Fluorophenoxy)piperidine

Precaución: for Research Use Only

Formula: C₁₁H₁₄FNO
MW: 195.2334 g/mol
MDL Number: MFCD04115015
CAS Number: 3413-28-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR