4,4,5,5-Tetramethyl-2-(triphenylen-2-yl)-1,3,2-dioxaborolane 95%

Supplier: Ambeed
Warning

Synonyms: Triphenylen-2-ylboronic acid Pinacol ester, 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)triphenylene

A639577-1G A639577-25G A639577-5G
77104-392EA 23.34 USD
77104-392 77104-394 77104-396
4,4,5,5-Tetramethyl-2-(triphenylen-2-yl)-1,3,2-dioxaborolane 95%
4,4,5,5-Tetramethyl-2-(triphenylen-2-yl)-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₂₄H₂₃BO₂
MW: 354.26 g/mol
MDL Number: MFCD26401844
CAS Number: 890042-13-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR