CPI-203 99+%

Supplier: Ambeed
Warning

Synonyms: (S)-2-(4-(4-Chlorophenyl)-2,3,9-trimethyl-6H-thieno[3,2-f][1,2,4]triazolo[4,3-a][1,4]diazepin-6-yl)acetamide

A639214-10MG A639214-1MG A639214-25MG A639214-2MG A639214-5MG
77104-382EA 54.11 USD
77104-382 77104-384 77104-386 77104-388 77104-390
CPI-203 99+%
CPI-203

Precaución: for Research Use Only

Formula: C₁₉H₁₈ClN₅OS
MW: 399.8971 g/mol
MDL Number: MFCD27997886
CAS Number: 1446144-04-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR