4-Fluoro-4'-nitrobenzophenone 98%

Supplier: Ambeed
Warning

Synonyms: (4-Fluorophenyl)(4-nitrophenyl)methanone

A639031-1G A639031-25G A639031-5G
77104-376EA 11.67 USD
77104-376 77104-378 77104-380
4-Fluoro-4'-nitrobenzophenone 98%
4-Fluoro-4'-nitrobenzophenone

Precaución: for Research Use Only

Formula: C₁₃H₈FNO₃
MW: 245.2059 g/mol
MDL Number: MFCD00053428
CAS Number: 2195-47-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR