2,4-Difluoro-3-isopropoxyphenylboronic acid 98%

Supplier: Ambeed
Warning

A638483-1G A638483-250MG A638483-5G
77104-370EA 128 USD
77104-370 77104-372 77104-374
2,4-Difluoro-3-isopropoxyphenylboronic acid 98%
2,4-Difluoro-3-isopropoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₁BF₂O₃
MDL Number: MFCD19981529
CAS Number: 1451390-95-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR