9-Acetyl-3,6-diiodocarbazole 95%

Supplier: Ambeed
Warning

Synonyms: 1-(3,6-Diiodo-9H-carbazol-9-yl)ethanone

A638017-250MG A638017-5G
77104-366EA 15 USD
77104-366 77104-368
9-Acetyl-3,6-diiodocarbazole 95%
9-Acetyl-3,6-diiodocarbazole

Precaución: for Research Use Only

Formula: C₁₄H₉I₂NO
MW: 461.04 g/mol
MDL Number: MFCD10000947
CAS Number: 606129-89-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR