Ethyl 4-chloro-1H-pyrazole-3-carboxylate 97+%

Supplier: Ambeed
Warning

A636780-100MG A636780-1G
77104-360EA 36.08 USD
77104-360 77104-362
Ethyl 4-chloro-1H-pyrazole-3-carboxylate 97+%
Ethyl 4-chloro-1H-pyrazole-3-carboxylate

Precaución: for Research Use Only

Formula: C₆H₇ClN₂O₂
MW: 174.58 g/mol
MDL Number: MFCD23378482
CAS Number: 1189363-78-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR