2-(3-Bromophenyl)-4,6-diphenylpyrimidine 98+%

Supplier: Ambeed
Warning

A636448-250MG A636448-5G A636448-1G
77104-354EA 11 USD
77104-354 77104-356 77686-583
2-(3-Bromophenyl)-4,6-diphenylpyrimidine 98+%
2-(3-Bromophenyl)-4,6-diphenylpyrimidine

Precaución: for Research Use Only

Formula: C₂₂H₁₅BrN₂
MDL Number: MFCD29477691
CAS Number: 864377-22-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR