1-(Trifluoroacetyl)piperidine 98%

Supplier: Ambeed
Danger

A635770-1G A635770-5G
77104-350EA 14 USD
77104-350 77104-352
1-(Trifluoroacetyl)piperidine 98%
1-(Trifluoroacetyl)piperidine

Precaución: for Research Use Only

Formula: C₇H₁₀F₃NO
MDL Number: MFCD00466213
CAS Number: 340-07-8
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR