(E)-Methyl 3-(4-(trifluoromethyl)phenyl)acrylate 98%

Supplier: Ambeed
Warning

A634316-10G A634316-1G A634316-5G
77104-332EA 145.35 USD
77104-332 77104-334 77104-336
(E)-Methyl 3-(4-(trifluoromethyl)phenyl)acrylate 98%
(E)-Methyl 3-(4-(trifluoromethyl)phenyl)acrylate

Precaución: for Research Use Only

Formula: C₁₁H₉F₃O₂
MW: 230.18 g/mol
MDL Number: MFCD01861883
CAS Number: 20754-22-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR