2-Hydroxy-9-fluorenone 96%

Supplier: Ambeed
Warning

Synonyms: 2-Hydroxy-9H-fluoren-9-one

A631727-1G A631727-250MG A631727-5G A631727-100MG
77104-320EA 111 USD
77104-320 77104-322 77104-324 77900-788
2-Hydroxy-9-fluorenone 96%
2-Hydroxy-9-fluorenone

Precaución: for Research Use Only

Formula: C₁₃H₈O₂
MDL Number: MFCD00001155
CAS Number: 6949-73-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR