Methyl 4-(benzylamino)-3-nitrobenzoate 96%

Supplier: Ambeed
Warning

A629087-1G A629087-5G
77104-292EA 29 USD
77104-292 77104-294
Methyl 4-(benzylamino)-3-nitrobenzoate 96%
Methyl 4-(benzylamino)-3-nitrobenzoate

Precaución: for Research Use Only

Formula: C₁₅H₁₄N₂O₄
MDL Number: MFCD09862452
CAS Number: 68502-46-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR