3,6-Dibromophthalic anhydride 98%

Supplier: Ambeed
Warning

Synonyms: 4,7-Dibromoisobenzofuran-1,3-dione

A628901-100MG A628901-1G A628901-250MG A628901-5G
77104-284EA 36.08 USD
77104-284 77104-286 77104-288 77104-290
3,6-Dibromophthalic anhydride 98%
3,6-Dibromophthalic anhydride

Precaución: for Research Use Only

Formula: C₈H₂Br₂O₃
MW: 305.91 g/mol
MDL Number: MFCD01117424
CAS Number: 25834-16-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR