(3-(Trifluoromethyl)pyridin-4-yl)boronic acid 95%

Supplier: Ambeed
Warning

A628831-100MG A628831-250MG
77104-280EA 200.52 USD
77104-280 77104-282
(3-(Trifluoromethyl)pyridin-4-yl)boronic acid 95%
(3-(Trifluoromethyl)pyridin-4-yl)boronic acid

Precaución: for Research Use Only

Formula: C₆H₅BF₃NO₂
MW: 190.92 g/mol
MDL Number: MFCD11975905
CAS Number: 1204334-17-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR