2,5-Dibromo-3-chloropyridine 98%

Supplier: Ambeed
Warning

Synonyms: 3-Chloro-2,5-dibromopyridine

A628494-10G A628494-1G A628494-250MG A628494-5G A628494-100G
77104-272EA 32 USD
77104-272 77104-274 77104-276 77104-278 77901-782
2,5-Dibromo-3-chloropyridine 98%
2,5-Dibromo-3-chloropyridine

Precaución: for Research Use Only

Formula: C₅H₂Br₂ClN
MW: 271.3371 g/mol
MDL Number: MFCD08277306
CAS Number: 160599-70-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR