N'-Hydroxypyridine-4-carboximidamide 98%

Supplier: Ambeed
Warning

Synonyms: N-Hydroxyisonicotinimidamide, 4-Pyridylamidoxime, Pyridine-4-amidoxime, N'-Hydroxy-4-pyridinecarboximidamide

A628257-1G A628257-25G A628257-5G
77104-266EA 10.61 USD
77104-266 77104-268 77104-270
N'-Hydroxypyridine-4-carboximidamide 98%
N'-Hydroxypyridine-4-carboximidamide

Precaución: for Research Use Only

Formula: C₆H₇N₃O
MW: 137.142 g/mol
MDL Number: MFCD00125873
CAS Number: 1594-57-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR