(R)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)pyrrolidine-3-carboxylic acid 95%

Supplier: Ambeed
Warning

A628117-1G A628117-250MG A628117-100MG A628117-5G
77104-264EA 88 USD
77104-264 77573-040 77900-673 77900-666
(R)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)pyrrolidine-3-carboxylic acid 95%
(R)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)pyrrolidine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₂₀H₁₉NO₄
MDL Number: MFCD14635841
CAS Number: 193693-65-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR