1,2-Di(4-boronophenoxy)ethane, dipinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 1,2-Bis(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenoxy)ethane, 1,2-Bis[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenoxy]ethane

A625075-100MG A625075-1G A625075-250MG
77104-258EA 36.08 USD
77104-258 77104-260 77104-262
1,2-Di(4-boronophenoxy)ethane, dipinacol ester 97%
1,2-Di(4-boronophenoxy)ethane, dipinacol ester

Precaución: for Research Use Only

Formula: C₂₆H₃₆B₂O₆
MW: 466.182 g/mol
MDL Number: MFCD09258655
CAS Number: 957061-07-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR