Benzofuran-2-carbonyl chloride 95%

Supplier: Ambeed
Danger

A624914-1G A624914-250MG A624914-100MG
77104-254EA 29 USD
77104-254 77104-256 77686-568
Benzofuran-2-carbonyl chloride 95%
Benzofuran-2-carbonyl chloride

Precaución: for Research Use Only

Formula: C₉H₅ClO₂
MDL Number: MFCD01659782
CAS Number: 41717-28-6
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR