2-Methoxy-5-(trifluoromethoxy)phenol 95+%

Supplier: Ambeed
Warning

A622927-1G A622927-250MG A622927-5G
77104-232EA 14 USD
77104-232 77104-234 77104-236
2-Methoxy-5-(trifluoromethoxy)phenol 95+%
2-Methoxy-5-(trifluoromethoxy)phenol

Precaución: for Research Use Only

Formula: C₈H₇F₃O₃
MDL Number: MFCD09832306
CAS Number: 895572-36-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR