[6-Methoxy-5-(trifluoromethyl)pyridin-3-yl]boronic acid 95%

Supplier: Ambeed
Warning

A621860-1G A621860-250MG
77104-230EA 100 USD
77104-230 77900-787
[6-Methoxy-5-(trifluoromethyl)pyridin-3-yl]boronic acid 95%
[6-Methoxy-5-(trifluoromethyl)pyridin-3-yl]boronic acid

Precaución: for Research Use Only

Formula: C₇H₇BF₃NO₃
MDL Number: MFCD18254530
CAS Number: 1420830-61-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR