Methyl 5-fluoro-2-methoxybenzoate 95%

Supplier: Ambeed
Warning

A620167-1G A620167-5G
77104-218EA 22 USD
77104-218 77104-220
Methyl 5-fluoro-2-methoxybenzoate 95%
Methyl 5-fluoro-2-methoxybenzoate

Precaución: for Research Use Only

Formula: C₉H₉FO₃
MDL Number: MFCD06203734
CAS Number: 151793-20-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR