2,2-Difluoro-2-(2-(trifluoromethoxy)phenyl)acetic acid 98%

Supplier: Ambeed
Danger

A619881-100MG A619881-1G A619881-250MG
77104-212EA 47.75 USD
77104-212 77104-214 77104-216
2,2-Difluoro-2-(2-(trifluoromethoxy)phenyl)acetic acid 98%
2,2-Difluoro-2-(2-(trifluoromethoxy)phenyl)acetic acid

Precaución: for Research Use Only

Formula: C₉H₅F₅O₃
MW: 256.1262 g/mol
MDL Number: MFCD11855928
CAS Number: 1133116-03-6
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR