α'-Bromo-α,α,α-trifluoro-o-xylene 98%

Supplier: Ambeed
Danger

A617003-25G A617003-5G A617003-10G
77104-202EA 25 USD
77104-202 77104-204 77569-168
α'-Bromo-α,α,α-trifluoro-o-xylene 98%
α'-Bromo-α,α,α-trifluoro-o-xylene

Precaución: For research use only.

Formula: C₈H₆BrF₃
MDL Number: MFCD00013558
CAS Number: 395-44-8
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR